ω-Bromo-4-chloro-3-nitroacetophenone, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Bromo-4-chloro-3-nitroacetophenone, 2-Bromo-4'-chloro-3'-nitroacetophenone, 4-Chloro-3-nitrophenacyl bromide

OR24997-10G OR24997-1G OR24997-25G
77407-578EA 309.72 USD
77407-578 77407-580 77407-582
ω-Bromo-4-chloro-3-nitroacetophenone, Technical Grade
ω-Bromo-4-chloro-3-nitroacetophenone
Formula: C₈H₅BrClNO₃
MW: 278.49
Melting Pt: 159…161 °C
Storage Temperature: Refrigerator
CAS Number: 22019-49-4
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR