4-Bromo-2-chlorobenzonitrile
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR48257-100G
OR48257-10G
OR48257-25G
77410-040EA
124.3
USD
77410-040
77410-042
77410-044
4-Bromo-2-chlorobenzonitrile
4-Bromo-2-chlorobenzonitrile
Formula:
C₇H₃BrClN Melting Pt: 67…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040883 CAS Number: 154607-01-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...