3-Bromo-2-chloro-4-methylpyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Bromo-2-chloro-4-picoline

OR301029-100G OR301029-25G OR301029-5G
77408-730EA 681.58 USD
77408-730 77408-732 77408-734
3-Bromo-2-chloro-4-methylpyridine 97%
3-Bromo-2-chloro-4-methylpyridine
Formula: C₆H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD04112511
CAS Number: 55404-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR