2,4-Dibromo-1H-imidazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR1229-1G OR1229-250MG
77404-244EA 124.3 USD
77404-244 77404-246
2,4-Dibromo-1H-imidazole 98%
2,4-Dibromo-1H-imidazole
Formula: C₃H₂Br₂N₂
MW: 225.87 g/mol
Melting Pt: 193…198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08741407
CAS Number: 64591-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR