4-Amino-4H-1,2,4-triazole 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR28852-100G
OR28852-25G
OR28852-500G
77408-268EA
23.4
USD
77408-268
77408-270
77408-272
4-Amino-4H-1,2,4-triazole 98%
4-Amino-4H-1,2,4-triazole
Formula:
C₂H₄N₄ Melting Pt: 83…88 °C Flash Pt: >149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003099 CAS Number: 584-13-4 EINECS: 209-533-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...