4-Fluoro-2-(trifluoromethyl)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Cyano-5-fluorobenzotrifluoride

PC4373J-100G PC4373J-25G PC4373J-500G
77415-282EA 65.2 USD
77415-282 77415-284 77415-286
4-Fluoro-2-(trifluoromethyl)benzonitrile 97%
4-Fluoro-2-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 194…196 °C
Melting Pt: 43…45 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00061283
CAS Number: 194853-86-6
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR