2-Fluoro-5-(trifluoromethyl)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC4373G-5G PC4373G-100G PC4373G-25G
77415-274EA 21.32 USD
77415-274 77415-270 77415-272
2-Fluoro-5-(trifluoromethyl)benzonitrile 97%
2-Fluoro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
Boiling Pt: 186 °C
Density: 1.38 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00061280
CAS Number: 4088-84-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR