4-Chloro-3-(trifluoromethyl)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Chloro-5-cyanobenzotrifluoride

PC2004-100G PC2004-25G PC2004-5G
77413-448EA 96.78 USD
77413-448 77413-450 77413-452
4-Chloro-3-(trifluoromethyl)benzonitrile 98%
4-Chloro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
Boiling Pt: 100…105 °C/11 mmHg
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00190117
CAS Number: 1735-54-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR