2-Chloro-4-(trifluoromethyl)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Chloro-4-cyanobenzotrifluoride

PC1713-100G PC1713-25G PC1713-5G
77413-278EA 179.88 USD
77413-278 77413-280 77413-282
2-Chloro-4-(trifluoromethyl)benzonitrile 98%
2-Chloro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
Boiling Pt: 192…193 °C
Density: 1.389 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00084939
CAS Number: 1813-33-8
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR