4,5-Dibromo-2-thenoic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4,5-Dibromo-2-thiophenecarboxylic acid, 4,5-Dibromothiophene-2-carboxylic acid

OR22419-25G OR22419-5G
77406-884EA 268.96 USD
77406-884 77406-886
4,5-Dibromo-2-thenoic acid
4,5-Dibromo-2-thenoic acid
Formula: C₅H₂Br₂O₂S
MW: 285.94 g/mol
Melting Pt: 225…232 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039682
CAS Number: 6324-10-3
EINECS: 228-683-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR