1-Bromo-3,3,3-trifluoro-2-propanol 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Bromo-1,1,1-trifluoropropan-2-ol, 3-Bromo-1,1,1-trifluoro-2-propanol

PC1599-1G PC1599-5G
77413-256EA 41.78 USD
77413-256 77413-258
1-Bromo-3,3,3-trifluoro-2-propanol 96%
1-Bromo-3,3,3-trifluoro-2-propanol

Precursor for 2-(Trifluoromethyl)oxirane.

Formula: BrH₂CCH(OH)CF₃
MW: 192.96 g/mol
Boiling Pt: 124…126 °C
Density: 1.861 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00040945
CAS Number: 431-34-5
UN: 1987
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR