4-(Methylsulphonyl)benzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Bromo-4-(methylsulphonyl)toluene, 1-(Bromomethyl)-4-(methylsulphonyl)benzene

OR46201-1G OR46201-5G
77409-894EA 28.52 USD
77409-894 77409-896
4-(Methylsulphonyl)benzyl bromide 97%
4-(Methylsulphonyl)benzyl bromide
Formula: C₈H₉BrO₂S
Melting Pt: 82…86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00185849
CAS Number: 53606-06-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR