2,3,4,5,6-Pentafluorostyrene 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC5810-25G PC5810-5G PC5810-100G
77417-114EA 150.22 USD
77417-114 77417-116 77417-112
2,3,4,5,6-Pentafluorostyrene 99%
2,3,4,5,6-Pentafluorostyrene
Formula: C₈H₃F₅
Boiling Pt: 139…140 °C
Density: 1.432 g/cm³ (25 °C)
Flash Pt: 34 °C
Storage Temperature: Freezer
MDL Number: MFCD00000300
CAS Number: 653-34-9
EINECS: 211-500-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR