5-Nitro-8-quinolinol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-Nitroquinolin-8-ol, Nitroxoline, 8-Hydroxy-5-nitroquinoline

OR5948-100G OR5948-25G
77410-602EA 110.04 USD
77410-602 77410-604
5-Nitro-8-quinolinol 98%
5-Nitro-8-quinolinol
Formula: C₉H₆N₂O₃
MW: 190.16 g/mol
Melting Pt: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00006791
CAS Number: 4008-48-4
EINECS: 223-662-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR