4-Fluorostyrene 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Fluoro-4-vinylbenzene

PC4250-25G PC4250-5G
77415-228EA 92.72 USD
77415-228 77415-230
4-Fluorostyrene 95%
4-Fluorostyrene
Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 145…148 °C
Melting Pt: –34.5 °C
Density: 1.024 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000361
CAS Number: 405-99-2
EINECS: 206-975-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR