2-Fluorostyrene 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Fluoro-2-vinylbenzene

PC4230-1G PC4230-25G PC4230-5G
77415-210EA 34.64 USD
77415-210 77415-212 77415-214
2-Fluorostyrene 99%
2-Fluorostyrene
Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 46 °C/32 mmHg
Density: 1.025 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013550
CAS Number: 394-46-7
EINECS: 206-896-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR