5-Amino-2-fluorobenzonitrile 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-Cyano-4-fluoroaniline

PC5823-100G PC5823-25G
77417-122EA 303.6 USD
77417-122 77417-124
5-Amino-2-fluorobenzonitrile 97%
5-Amino-2-fluorobenzonitrile
Formula: C₇H₅FN₂
MDL Number: MFCD00277872
CAS Number: 53312-81-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR