Heptafluorobutyric anhydride (HFAA) 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: HFBA, 2,2,3,3,4,4,4-Heptafluorobutanoic anhydride, Perfluorobutyric anhydride, HFAA

PC4470-100G PC4470-25G
77415-434EA 282.22 USD
77415-434 77415-436
Heptafluorobutyric anhydride (HFAA) 95%
Heptafluorobutyric anhydride

Useful for derivitisation of amines, alcohols, amino acids and other bioactive compounds for GC analysis.

Formula: C₈F₁₄O₃
Boiling Pt: 106…107 °C
Melting Pt: –43 °C
Density: 1.665 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000432
CAS Number: 336-59-4
EINECS: 206-410-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR