2-Fluoro-6-hydroxybenzonitrile 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC10435-1G PC10435-25G PC10435-5G
77412-910EA 21.26 USD
77412-910 77412-912 77412-914
2-Fluoro-6-hydroxybenzonitrile 99%
2-Fluoro-6-hydroxybenzonitrile
Formula: C₇H₄FNO
Melting Pt: 154…158 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03428592
CAS Number: 140675-43-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR