2-Amino-6-fluorobenzonitrile 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC1071Q-100G
PC1071Q-500G
77412-964EA
33.62
USD
77412-964
77412-966
2-Amino-6-fluorobenzonitrile 98%
2-Amino-6-fluorobenzonitrile
Formula:
C₇H₅FN₂ Melting Pt: 125…129 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00015477 CAS Number: 77326-36-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...