Cytosine 98%

Supplier: APOLLO SCIENTIFIC

OR18333-500G OR18333-100G
77406-234EA 75.19 USD
77406-234 77406-236 77406-232
Cytosine 98%
Cytosine
Formula: C₄H₅N₃O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006034
CAS Number: 71-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR