2-(Bromomethyl)phenylboronic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Bromomethyl)benzeneboronic acid, Dihydroxy(2-(bromomethyl)phenyl)borane, (2-(Bromomethyl)phenyl)metaboric acid, 2-Bromomethylphenylboronic acid

OR10377-1G OR10377-5G
77402-656EA 21.26 USD
77402-656 77402-658
2-(Bromomethyl)phenylboronic acid 95%
2-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
Melting Pt: 145…148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318941
CAS Number: 91983-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR