Tris(4-methoxyphenyl)phosphine 95%
Supplier: APOLLO SCIENTIFIC
Synonyms:
Tris(p-methoxyphenyl)phosphine
OR315480-1G
OR315480-25G
OR315480-5G
77409-070EA
21.26
USD
77409-070
77409-072
77409-074
Tris(4-methoxyphenyl)phosphine 95%
Tris(4-methoxyphenyl)phosphine
Formula:
C₂₁H₂₁O₃P Melting Pt: 131…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014896 CAS Number: 855-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...