Tetrachlorophthalic anhydride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR460073-100G OR460073-500G
77409-864EA 48.9 USD
77409-864 77409-866
Tetrachlorophthalic anhydride 97%
Tetrachlorophthalic anhydride
Formula: C₈Cl₄O₃
MW: 285.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005920
CAS Number: 117-08-8
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR