Isoxazole-5-carbonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR2811-1G OR2811-250MG OR2811-5G
77408-030EA 44.34 USD
77408-030 77408-032 77408-034
Isoxazole-5-carbonyl chloride 97%
Isoxazole-5-carbonyl chloride
Formula: C₄H₂ClNO₂
Boiling Pt: 167…168 °C
Density: 1.362 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00067868
CAS Number: 62348-13-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR