4-Methoxybenzenesulphonyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Methoxybenzene-1-sulphonyl chloride

OR30190-100G OR30190-25G
77408-820EA 49.92 USD
77408-820 77408-822
4-Methoxybenzenesulphonyl chloride 98%
4-Methoxybenzenesulphonyl chloride
Formula: C₇H₇ClO₃S
Boiling Pt: 119…122 °C/0.4 mmHg
Melting Pt: 39…43 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007446
CAS Number: 98-68-0
EINECS: 202-692-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR