4-Bromo-2,6-dichlorobenzenesulfonyl chloride 96%

Supplier: APOLLO SCIENTIFIC
Danger

OR4495-1G OR4495-25G OR4495-5G
77409-742EA 21.26 USD
77409-742 77409-744 77409-746
4-Bromo-2,6-dichlorobenzenesulfonyl chloride 96%
4-Bromo-2,6-dichlorobenzenesulphonyl chloride
Formula: C₆H₂BrCl₃O₂S
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094651
CAS Number: 351003-54-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR