3,3,3-Trifluoropropionic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3,3,3-Trifluoropropanoic acid

PC7820A-100G PC7820A-25G PC7820A-5G
77418-234EA 261.84 USD
77418-234 77418-236 77418-238
3,3,3-Trifluoropropionic acid 98%
3,3,3-Trifluoropropionic acid
Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 146 °C
Melting Pt: 9…10 °C
Density: 1.441 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00153292
CAS Number: 2516-99-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR