Isobutyric anhydride 95%
Supplier: APOLLO SCIENTIFIC
Synonyms:
2-Methylpropanoic anhydride, 2-Methylpropionic anhydride
OR923617-100G
OR923617-500G
77411-868EA
117.16
USD
77411-868
77411-870
Isobutyric anhydride 95%
Isobutyric anhydride
Formula:
C₈H₁₄O₃ MW: 158.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008913 CAS Number: 97-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...