3-Fluoropyridine-2-carbonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1215-100G PC1215-25G PC1215-5G
77413-080EA 205.8 USD
77413-080 77413-082 77413-084
3-Fluoropyridine-2-carbonitrile 97%
3-Fluoropyridine-2-carbonitrile
Formula: C₆H₃FN₂
Boiling Pt: 120…125 °C/17 mmHg
Melting Pt: 27…32 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD06797501
CAS Number: 97509-75-6
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR