2-Chloro-6-(trifluoromethyl)pyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC4544-100G PC4544-25G PC4544-500G
77415-616EA 69.28 USD
77415-616 77415-618 77415-620
2-Chloro-6-(trifluoromethyl)pyridine 98%
2-Chloro-6-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
MW: 181.54
Boiling Pt: 60…62 °C/8 mmHg
Melting Pt: 32…36 °C
Flash Pt: 79.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00042226
CAS Number: 39890-95-4
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR