2-Bromo-6-(trifluoromethyl)pyridine 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC7657-100G
PC7657-25G
PC7657-5G
77418-086EA
349.46
USD
77418-086
77418-088
77418-090
2-Bromo-6-(trifluoromethyl)pyridine 98%
2-Bromo-6-(trifluoromethyl)pyridine
Formula:
C₆H₃BrF₃N MW: 226 Boiling Pt: 78…79 °C/14 mmHg Melting Pt: 48…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153087 CAS Number: 189278-27-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...