α-Bromo-o-toluic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR16987-1G OR16987-25G OR16987-5G
77405-880EA 55.02 USD
77405-880 77405-882 77405-884
α-Bromo-o-toluic acid 97%
α-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
Melting Pt: 145…150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00090420
CAS Number: 7115-89-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR