α,α,α-Trifluoro-m-xylene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC4507-1G PC4507-25G PC4507-5G
77415-594EA 21.26 USD
77415-594 77415-596 77415-598
α,α,α-Trifluoro-m-xylene 98%
α,α,α-Trifluoro-m-xylene
Formula: C₈H₇F₃
Boiling Pt: 130 °C
Density: 1.148 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00190105
CAS Number: 401-79-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR