4-Fluoro-3-(trifluoromethyl)phenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Fluoro-5-hydroxybenzotrifluoride, 4-Fluoro-3-trifluoromethylphenol

PC4376R-25G PC4376R-5G PC4376R-100G
77415-374EA 43.8 USD
77415-374 77415-376 77415-372
4-Fluoro-3-(trifluoromethyl)phenol 98%
4-Fluoro-3-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 84…86 °C/15 mmHg
Melting Pt: 17 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00043875
CAS Number: 61721-07-1
EINECS: 432-560-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR