α,α,α-Trifluoro-m-toluonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7560-100G PC7560-25G
77418-004EA 48.9 USD
77418-004 77418-006
α,α,α-Trifluoro-m-toluonitrile 98%
α,α,α-Trifluoro-m-toluonitrile
Formula: C₈H₄F₃N
Boiling Pt: 189 °C
Melting Pt: 14.5 °C
Density: 1.2821 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00001807
CAS Number: 368-77-4
EINECS: 206-711-4
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR