2-Chloroethyl methyl ether

Supplier: APOLLO SCIENTIFIC
Danger

OR17824-100G OR17824-25G OR17824-500G
77406-106EA 37.7 USD
77406-106 77406-108 77406-110
2-Chloroethyl methyl ether
2-Chloroethyl methyl ether
Formula: C₃H₇ClO
Boiling Pt: 89…90 °C
Melting Pt: –55 °C
Density: 1.035 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000972
CAS Number: 627-42-9
EINECS: 210-999-7
UN: 3271
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR