4-Bromobenzhydrol 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: p-Bromobenzhydrol, 4-Bromophenyl phenyl carbinol, 4-Bromodiphenylmethanol, 4-Bromo-α-phenylbenzyl alcohol, 4-Bromobenzhydryl alcohol, (4-Bromophenyl)(phenyl)methanol

OR4692-1G OR4692-25G OR4692-5G
77409-954EA 21.4 USD
77409-954 77409-956 77409-958
4-Bromobenzhydrol 96%
4-Bromobenzhydrol
Formula: C₁₃H₁₁BrO
MW: 263.13
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00016851
CAS Number: 29334-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR