1-Bromo-5-chloro-2-iodobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11085-100G OR11085-25G OR11085-5G
77403-514EA 121.24 USD
77403-514 77403-520 77403-522
1-Bromo-5-chloro-2-iodobenzene 98%
1-Bromo-5-chloro-2-iodobenzene
Formula: C₆H₃BrClI
Storage Temperature: Ambient
MDL Number: MFCD00672944
CAS Number: 31928-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR