2-Bromo-5-(trifluoromethyl)benzyl bromide 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
4-Bromo-3-(bromomethyl)benzotrifluoride, 1-Bromo-2-(bromomethyl)-4-(trifluoromethyl)benzene
PC7887-10G
PC7887-1G
PC7887-5G
77418-280EA
211.92
USD
77418-280
77418-282
77418-284
2-Bromo-5-(trifluoromethyl)benzyl bromide 97%
2-Bromo-5-(trifluoromethyl)benzyl bromide
Formula:
C₈H₅Br₂F₃ MW: 317.93 Storage Temperature: Ambient |
MDL Number:
MFCD04973762 CAS Number: 886496-63-5 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...