2-Nitro-4-(trifluoromethoxy)aniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC5292-100G PC5292-25G PC5292-5G
77416-584EA 84.32 USD
77416-584 77416-586 77416-588
2-Nitro-4-(trifluoromethoxy)aniline 98%
2-Nitro-4-(trifluoromethoxy)aniline
Formula: C₇H₅F₃N₂O₃
Melting Pt: 62…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042326
CAS Number: 2267-23-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR