N-Phenyl-o-phenylenediamine 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: N-(2-Aminophenyl)aniline, 2-Aminodiphenylamine, N1-Phenylbenzene-1,2-diamine

OR6646-100G OR6646-10G OR6646-25G
77410-884EA 84.56 USD
77410-884 77410-886 77410-888
N-Phenyl-o-phenylenediamine 98%
N-Phenyl-o-phenylenediamine
Formula: C₁₂H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00007685
CAS Number: 534-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR