3-Methylpyrocatechol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Methylcatechol, 3-Methylbenzene-1,2-diol

OR28777-100G OR28777-25G OR28777-5G
77408-222EA 308.7 USD
77408-222 77408-224 77408-226
3-Methylpyrocatechol 98%
3-Methylpyrocatechol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 241 °C
Melting Pt: 65…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00016435
CAS Number: 488-17-5
EINECS: 207-672-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR