α,α,α-Trifluoro-3,4-diaminotoluene 97+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Trifluoromethyl)-1,2-phenylenediamine, 4-(Trifluoromethyl)benzene-1,2-diamine, 3,4-Diaminobenzotrifluoride, 4-(Trifluoromethyl)-o-phenylenediamine

PC2168-100G PC2168-25G PC2168-5G
77413-550EA 206.82 USD
77413-550 77413-552 77413-554
α,α,α-Trifluoro-3,4-diaminotoluene 97+%
α,α,α-Trifluoro-3,4-diaminotoluene
Formula: C₇H₇F₃N₂
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00042456
CAS Number: 368-71-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR