4-Fluoro-3-(trifluoromethyl)benzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Bromomethyl)-1-fluoro-2-(trifluoromethyl)benzene, 1-(Bromomethyl)-4-fluoro-3-(trifluoromethyl)benzene

PC4376K-100G PC4376K-25G PC4376K-5G
77415-356EA 177.28 USD
77415-356 77415-358 77415-360
4-Fluoro-3-(trifluoromethyl)benzyl bromide 98%
4-Fluoro-3-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
MW: 257.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061177
CAS Number: 184970-26-1
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR