3,5-Bis(trifluoromethyl)benzyl bromide 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene
PC3105-100G
PC3105-25G
PC3105-500G
77414-480EA
124.3
USD
77414-480
77414-482
77414-484
3,5-Bis(trifluoromethyl)benzyl bromide 98%
3,5-Bis(trifluoromethyl)benzyl bromide
Formula:
C₉H₅BrF₆ MW: 307.03 g/mol Boiling Pt: 136…140 °C/14 mmHg Melting Pt: 18 °C Density: 1.675 g/cm³ (25 °C) Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009905 CAS Number: 32247-96-4 EINECS: 250-971-1 UN: 2920 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...