2-Fluoro-5-(trifluoromethyl)benzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-2-fluoro-5-(trifluoromethyl)benzene, 2-Fluoro-5-(trifluoromethyl)benzyl Bromide

PC4376G-1G PC4376G-25G PC4376G-5G
77415-344EA 21.26 USD
77415-344 77415-346 77415-348
2-Fluoro-5-(trifluoromethyl)benzyl bromide 97%
2-Fluoro-5-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
Storage Temperature: Ambient
MDL Number: MFCD00061174
CAS Number: 220239-69-0
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR