4-Fluoro-2-(trifluoromethyl)benzylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (4-Fluoro-2-(trifluoromethyl)phenyl)methanamine

PC4374Y-100G PC4374Y-25G PC4374Y-5G
77415-326EA 430.08 USD
77415-326 77415-328 77415-330
4-Fluoro-2-(trifluoromethyl)benzylamine 97%
4-Fluoro-2-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
Storage Temperature: Ambient
MDL Number: MFCD00061164
CAS Number: 202522-22-3
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR