4-Chloro-3-fluorobenzylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC302780-10G PC302780-1G PC302780-5G
77414-258EA 81.5 USD
77414-258 77414-260 77414-262
4-Chloro-3-fluorobenzylamine 97%
4-Chloro-3-fluorobenzylamine
Formula: C₇H₇ClFN
Storage Temperature: Ambient
MDL Number: MFCD04116008
CAS Number: 72235-58-6
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR