5-Fluoro-2-(trifluoromethyl)benzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC0354-10G PC0354-1G PC0354-5G
77412-502EA 230.26 USD
77412-502 77412-504 77412-506
5-Fluoro-2-(trifluoromethyl)benzylamine 98%
5-Fluoro-2-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
MW: 193.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236288
CAS Number: 231291-14-8
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR